missing translation for 'onlineSavingsMsg'
Learn More

Dulcin (>90%), TRC

Product Code. 30009988
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009988 25 g 25g
30004546 10 g 10g
17952965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009988 Supplier Toronto Research Chemicals Supplier No. TRCD72045025G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-69-6
Molecular Formula C9 H12 N2 O2
Molecular Weight (g/mol) 180.2
Synonym Urea, (4-ethoxyphenyl)- (9CI), Urea, (p-ethoxyphenyl)- (8CI), N-(4-Ethoxyphenyl)urea, (4-Ethoxyphenyl)urea, (p-Ethoxyphenyl)urea, 1-(4-Ethoxyphenyl)urea, 4-Ureidophenetole, Dulcin, Dulcin (sweetener), Dulcine, NSC 1839, Phenetolcarbamide, Sucrol, Valzin, p-Phenetolcarbamide, p-Phenetylurea
IUPAC Name (4-ethoxyphenyl)urea
SMILES CCOc1ccc(NC(=O)N)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H12N2O2/c1-2-13-8-5-3-7(4-6-8)11-9(10)12/h3-6H,2H2,1H3,(H3,10,11,12)
Formula Weight 180.0899
Chemical Name or Material Glycosine (Dulcin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.