Learn More
MP Biomedicals Dulbecco's Modified Eagle's Medium (DMEM) without L-Glutamine and Phenol Red
Shop All MP Biomedicals ProductsDescription
Contains four times greater concentration of amino acids, vitamins and supplementary components.
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures. For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available from MP.
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications: Mammalian Cell Culture
Specifications
Specifications
| Concentration | 1 X |
| Form | Solution |
| Product Type | Eagle, Dulbecco Modified Eagle, DMEM |
| Quantity | 500 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.