missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Dulbecco's Modification of Eagle's Medium with HEPES and 4.5 g Dextrose/L, without L-Glutamine or Sodium Bicarbonate

Product Code. p-8015240 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
3 L
500mL
This item is not returnable. View return policy

Product Code. 11470355

Brand: MP Biomedicals™ 091233454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for supporting and maintaining a wide range of mammalian cell types.

Dulbecco's Modified Eagle's Medium (DMEM) is the most widely used modification of Eagle's Basal Medium (BME). DMEM contains four times greater concentration of amino acids, vitamins and supplementary components.

Also known as DME, Dulbecco's Modified Eagle's Medium is the most widely used modification of Eagle's Basal Medium (BME).

DMEM contains four times greater concentration of amino acids, vitamins and supplementary components. The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.

For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available from MP.

Dulbecco's Modification of Eagle's Medium with 20 mM HEPES and 4.5 g Dextrose/liter, without L-glutamine or Sodium Bicarbonate is ideally suited for supporting and maintaining a vast range of mammalian cell types.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Cell Culture Media
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Dextrose, HEPES
Without Additives No Glutamine, No Sodium Bicarbonate
For Use With (Application) Mammalian Cell Culture
Quantity 500 mL
pH 7.2 to 7.6
Content And Storage Store at 4°C
Description DMEM
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.