Learn More
LTE Scientific™ Dual Steam Option for Touchclave K Model Autoclaves
Click to view available options
For Use With (Equipment):
All Touchclave K Models
Description
- Dual Steam Option
- Allows for direct steam supply to be fitted where available
- Machine fitted with integral steam generator
- Dual operation provides additional peace of mind
Specifications
Specifications
| For Use With (Equipment) | All Touchclave K Models |
| Model | TC/DSO/01 |
| Sterility | Non-sterile |