Learn More
VEE GEE Dual Scale Hydrometer
The Dual Scale Hydrometer measures both heavy and light liquids with separate scales for each. It is calibrated for use at 60°F.
Brand:  VEE GEE 6603DS-2
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 17856623
  • £29.65 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • The Dual Scale Specific Gravity/Baume Hydrometer includes two scales.
  • One scale is designed for liquids heavier than water.
  • The other scale is specifically for liquids lighter than water.
Specifications
Specifications
Specific Gravity and Baume Hydrometer
30.5 cm
1.200 to 1.420
0.002/0.2°
25° to 42° (Heavy)
15.6°C (60°F)