missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Dual Plate Holder Assembly for FP-96

Product Code. 15884185 Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-96
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15884185

Brand: MP Biomedicals 119696168

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with FastPrep-96

  • For use with FastPrep-96
  • Ambient type adapter
  • High sample throughput
  • 96-well plate tube compatibility
TRUSTED_SUSTAINABILITY

Specifications

Description Sample throughput: High
For Use With (Equipment) FastPrep-96
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.