missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dual-Light™ Luciferase & β-Galactosidase Reporter Gene Assay System

Product Code. 12664265
Click to view available options
Quantity:
200 Assays
4000 Assays
600 Assays
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12664265

Brand: Invitrogen™ T1005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Enzyme substrate, lysis solution, required buffers, and reaction accelerator

Rapid and sensitive detection of firefly luciferase and E. coli β-galactosidase in a single extract aliquot for gene expression assays.

  • Detection of firefly luciferase and β-galactosidase in same sample simplifies normalization of transfection efficiency
  • Wide dynamic range of β-galactosidase and luciferase assays enables accurate measurement of both enzymes from femtogram to nanogram range
  • Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround

Easy-to-Use
Light signal from each enzymatic reaction is measured sequentially in a luminometer with automatic injectors or other instrumentation in which light emission measurements can be performed within a short period. First, luciferase reporter enzyme activity is quantitated with an enhanced luciferase reaction. Following a 30-60 minute incubation and addition of a light-emission accelerator, β-galactosidase reporter enzyme activity is determined with Galacton-Plus™ substrate. Both assays are combined into one simple assay using only one extract aliquot for greater convenience and precision. The entire assay is completed in less than one hour.

Wide Dynamic Range
The wide dynamic range of this dual assay enables accurate measurement of luciferase and β-galactosidase concentrations over seven orders of magnitude, from the femtogram to nanogram range. Colorimetric and isotopic reporter gene assays cannot rival the dynamic range of the Dual-Light system. We have formatted this assay for tube or microplate luminometer, with injection capability.

Specifications

  • Compatible Cells: Mammalian Cells, Yeast Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate), Luminometer (Tube-Based)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Galacton-Plus, Luciferin
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Beta-Gal Substrate, Luciferase Substrate
TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase/Luciferase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate), Luminometer (Tube-Based)
Assay β-Gal (lacZ) Assay, Luciferase Assay
Content And Storage This kit contains the enzyme substrate, lysis solution, required buffers, and reaction acccelerator.
Store the product at -20°C.
Compatible Cells Mammalian Cells, Yeast Cells
High-throughput Compatibility High-throughput Compatible
Readout End Point
Sample Type Cell Cultures
Substrate Galacton-Plus™, Luciferin
Substrate Type Beta-Gal Substrate, Luciferase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Luciferase, Beta-Galactosidase
Product Line Dual-Light™, NovaBright™
For Use With (Application) Enzyme Assay
Quantity 600 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.