missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dU Phosphoramidite, standard grade, serum vial bottle

Product Code. 18044090 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μM
1 g
Unit Size:
100µmol
1g
This item is not returnable. View return policy

Product Code. 18044090

Brand: Thermo Scientific™ 27173803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis.

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C39H47N4O8P
Base: dU
Formula weight: 730.8 g/mol
CAS number: 109389-30-2
HPLC Purity: ≥98.0%; conforms to reference
31P NMR Purity: ≥98.0%; conforms to reference

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type dU CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyuridine (dU)
Shipping Condition Room Temperature
Purity 98%
Quantity 100 μM
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.