missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DTX3 Monoclonal Antibody (4F10)
GREENER_CHOICE

Product Code. 15652225
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15652225 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652225 Supplier Invitrogen™ Supplier No. MA524397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MCGRFYGQLV GNQPQNGRML VSKDATLLLP SYEKYGTIVI QYVFPPGVQG AEHPNPGVRY PGTTRVAYLP DCPEGNKVLT LFRKAFDQRL TFTIGTSMTT.

Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX3
Applications ELISA
Classification Monoclonal
Clone 4F10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene DTX3
Gene Accession No. Q8N9I9
Gene Alias deltex 3 homolog (Drosophila); deltex 3, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 3; deltex homolog 3; deltex3; DTX3; FLJ34766; mDTX3; mg310; MGC138863; MGC138864; probable E3 ubiquitin-protein ligase DTX3; Protein deltex-3; RING finger protein 154; RING-type E3 ubiquitin transferase DTX3; RNF154
Gene Symbols DTX3
Host Species Mouse
Immunogen DTX3 (NP_848597, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196403
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.