missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-16030-100UL
This item is not returnable.
View return policy
Description
DTX1 Polyclonal antibody specifically detects DTX1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DTX1 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| deltex homolog 1 (Drosophila), deltex1, hDTX1, hDx-1, protein deltex-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human DTX1 (NP_004407.2). | |
| 100 μg | |
| Signal Transduction | |
| 1840 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction