missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584249
This item is not returnable.
View return policy
Description
Immunogen sequence: PPVSKSDVKP VPGVPGVCRK TKKKHLKKSK NPEDVVRRYM QKVKNPP Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.
Specifications
| DTX1 | |
| Polyclonal | |
| Unconjugated | |
| Dtx1 | |
| deltex 1; deltex 1 homolog (Drosophila); deltex 1, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 1; deltex homolog 1; deltex1; DTX1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; Fxit1; FXI-T1; hDTX1; hDx-1; mDTX1; mKIAA4160; protein deltex-1; RING-type E3 ubiquitin transferase DTX1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y01 | |
| Dtx1 | |
| Recombinant protein corresponding to Human DTX1. Recombinant protein control fragment (Product #RP-105011). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction