missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX1/DTX4 Antibody (730612) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7157JF525
This item is not returnable.
View return policy
Description
DTX1/DTX4 Monoclonal antibody specifically detects DTX1/DTX4 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| DTX1/DTX4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| DTX1;Fxit1;hDx-1;mKIAA4160 | |
| E. coli-derived recombinant human DTX1/DTX4, Met1-Phe147, Accession # Q86Y01 | |
| RUO | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 730612 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction