missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTNBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110982
This item is not returnable.
View return policy
Description
Immunogen sequence: GLKTLSDKSR EAKVKSKPRT VPFLPKYSAG LELLSRYEDT WAALHRRAKD CASAGELVDS EVVMLSAHWE KK Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 90%.
Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
Specifications
| DTNBP1 | |
| Polyclonal | |
| Unconjugated | |
| DTNBP1 | |
| 5430437B18Rik; AW048963; biogenesis of lysosomal organelles complex-1, subunit 8; biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; Bloc1s8; DBND; distrobrevin binding protein 1; DKFZp564K192; DTBP1; Dtnbp1; Dysbindin; dysbindin-1; dystrobrevin binding protein 1; dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; hermansky-Pudlak syndrome 7 protein homolog; HPS7; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; My031; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy; sdy | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EV8 | |
| DTNBP1 | |
| Recombinant Protein corresonding to Human DTNBP1. Recombinant protein control fragment (Product #RP-94606). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction