missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTL Polyclonal Antibody

Description
DTL Polyclonal Antibody for Western Blot, Flow
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control, DNA damage response and translesion DNA synthesis. The DCX(DTL) complex, also named CRL4(CDT2) complex, mediates the polyubiquitination and subsequent degradation of CDT1 and CDKN1A/p21(CIP1). CDT1 degradation in response to DNA damage is necessary to ensure proper cell cycle regulation of DNA replication. CDKN1A/p21(CIP1) degradation during S phase or following UV irradiation is essential to control replication licensing. Most substrates require their interaction with PCNA for their polyubiquitination: substrates interact with PCNA via their PIP-box, and those containing the 'K+4' motif in the PIP box, recruit the DCX(DTL) complex, leading to their degradation. In undamaged proliferating cells, the DCX(DTL) complex also promotes the 'Lys-164' monoubiquitination of PCNA, thereby being involved in PCNA-dependent translesion DNA synthesis.
Specifications
Specifications
| Antigen | DTL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DTL |
| Gene Accession No. | Q3TLR7, Q9NZJ0 |
| Gene Alias | 2810047L02Rik; 5730564G15Rik; CDT2; CDW1; DCAF2; DDB1 and CUL4 associated factor 2; DDB1- and CUL4-associated factor 2; denticleless E3 ubiquitin protein ligase homolog; denticleless E3 ubiquitin protein ligase homolog (Drosophila); denticleless homolog; denticleless homolog (Drosophila); denticleless protein homolog; DTL; L2dtl; lethal(2) denticleless protein homolog; Meth A RAMP; Meth A retinoic acid-regulated nuclear matrix-associated protein; RA regulated nuclear matrix associated protein; RAMP; RA-regulated nuclear matrix-associated protein; Retinoic acid-regulated nuclear matrix-associated protein; RGD1310439 |
| Gene Symbols | DTL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?