missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dtk/TYRO3 Antibody, PE/Atto594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28635PEATT594
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| Dtk/TYRO3 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular) | |
| Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
| 7301.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| TYRO3 | |
| Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot No. Q06418] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction