missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dtk/TYRO3 Antibody (ELB031) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28751JF669
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| TYRO3 | |
| 0.1 mL | |
| Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
| 7301 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ELB031 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction