missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dtk/TYRO3 Antibody (ELB031) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Dtk/TYRO3 |
|---|---|
| Clone | ELB031 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044387
|
Novus Biologicals™
NBP3-28751-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044219
|
Novus Biologicals™
NBP3-28751-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044236
|
Novus Biologicals™
NBP3-28751-100ug |
100 μg |
£256.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Dtk/TYRO3 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| TYRO3 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| ELB031 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q06418 | |
| 7301 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title