missing translation for 'onlineSavingsMsg'
Learn More

DTG, TRC

Product Code. 30380051
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380051 5 g 5g
30378349 10 g -
30376357 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380051 Supplier Toronto Research Chemicals Supplier No. TRCD7100105G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-39-2
Molecular Formula C15 H17 N3
Molecular Weight (g/mol) 239.32
Synonym 1,3-Bis(2-o-tolyl)guanidine, Guanidine, 1,3-di-o-tolyl- (6CI,7CI,8CI), N,N'-Bis(2-methylphenyl)guanidine, 1,2-Bis(2-methylphenyl)guanidine, 1,3-Bis(o-tolyl)guanidine, 1,3-Di-2-tolylguanidine, 1,3-Di-o-tolylguanidine, 1,3-Di-ortho-tolylguanidine, Akrochem DOTG, CNS 1001, DOTG, DTG, Di-o-tolylguanidine, Eveite DOTG, N,N'-Di-o-toluylguanidine, N,N'-Di-o-tolylguanidine, NSC 132023, NSC 473, Nocceler DT, Rhenogran DOTG 70, Sanceler DT, Soxinol DT, Vulcafor DOTG, Vulkacit DOTG
IUPAC Name 1,3-bis(2-methylphenyl)guanidine
SMILES Cc1ccccc1NC(=N)Nc2ccccc2C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H17N3/c1-11-7-3-5-9-13(11)17-15(16)18-14-10-6-4-8-12(14)2/h3-10H,1-2H3,(H3,16,17,18)
Formula Weight 239.1422
Chemical Name or Material 1,3-Bis(2-o-tolyl)guanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.