missing translation for 'onlineSavingsMsg'
Learn More

DTE, MedChemExpress

Product Code. 16499447
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499447 5 g 5g
16491569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499447 Supplier Medchem Express Supplier No. HY15916
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DTE (Dithioerythritol) is a sulfur containing sugar derived from the corresponding 4-carbon monosaccharide erythrose; is an epimer of dithiothreitol(DTT).

DTE (Dithioerythritol) is a sulfur containing sugar derived from the corresponding 4-carbon monosaccharide erythrose

Specifications

Chemical Name or Material DTE
CAS 6892-68-8
Color White
Purity Grade Notes Research
Molecular Formula C4H10O2S2
Quantity 5 g
Synonym Dithioerythritol Cleland’s reagent
Solubility Information DMSO : 100 mg/mL (648.30 mM; Need ultrasonic) ∣H2O : 50 mg/mL (324.15 mM; Need ultrasonic)
SMILES O[C@H]([C@H](O)CS)CS
Molecular Weight (g/mol) 154.25
Formula Weight 154.25
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.