Learn More
3M™ DT-Series Combination Filter
Product Code. 17796973
Change view
Click to view available options
Model:
CF22 A2B2E1P3 R D, DT-4036E
CF22 A2B2P3 R D, DT-4035E
CF22 B2P3 R D, DT-4032E
CF32 A2B2E2K2HgP3 R D, DT-4031E
CF32 A2B2E2K2HgP3 R D, DT-4046E
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17796973 CF22 A2B2E1P3 R D, DT-4036E Pack of 10
17706983 CF22 B2P3 R D, DT-4032E Pack of 10
17716983 CF22 A2B2P3 R D, DT-4035E Pack of 10
17726983 CF32 A2B2E2K2HgP3 R D, DT-4046E Pack of 10
17736983 CF32 A2B2E2K2HgP3 R D, DT-4031E Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17796973 Supplier 3M™ Supplier No. 7100260627
DT series filters are used for effective protection against a range of particulates, gases, vapours, or combination hazards.
  • Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
  • Features a DIN40 mm threaded connection
  • Please note DT-Series filters were previously known as Pro2000 Filters
  • To identify which filter is suitable for your environment, and to aid in the implementation of a filter changeout schedule, please use the 3M Service Life and Select Software, available from www.3m.com/sls.
TRUSTED_SUSTAINABILITY
Specifications
Cartridge/Filter COMBINATION
Filter Type Combination Organic Vapor/Acid Gas/HEPA
Product Type Filter
For Use With (Equipment) 3M™ Air Purifying Respirators, 3M™ PAPR Systems
Model CF22 A2B2E1P3 R D, DT-4036E
Quantity 10/Cs.