missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSS1 Polyclonal Antibody

DSS1 Polyclonal Antibody
Brand: Thermo Scientific PA5106312
This item is not returnable.
View return policy
Description
DSS1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| DSS1 | |
| Polyclonal | |
| Unconjugated | |
| SEM1 | |
| 26S proteasome complex subunit DSS1; 26S proteasome complex subunit SEM1; C7orf76; Deleted in split hand/split foot protein 1; deleted in split hand/split foot protein 1 homolog; deleted in split-hand/split-foot 1; Dss1; ECD; Putative protein SEM1, isoform 2; Sem1; SHFD1; Shfdg1; Shfg; Shfm1; SHSF1; split hand/foot deleted gene 1; split hand/foot deleted protein 1; Split hand/foot deleted protein 1 homolog; split hand/foot malformation (ectrodactyly) type 1; split hand/foot malformation type 1 protein; Split hand/foot malformation type 1 protein homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 0, 20422, 680532, 7979 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60896, P60897, Q6ZVN7 | |
| SEM1 | |
| A synthesized peptide derived from human SEM1(Accession P60896), corresponding to amino acid residues D35-R57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction