missing translation for 'onlineSavingsMsg'
Learn More

DSS1 Antibody, Novus Biologicals™

Product Code. 18322506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322506 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322506 Supplier Novus Biologicals Supplier No. NBP303876

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DSS1 Polyclonal specifically detects DSS1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 26S proteasome complex subunit DSS1, Deleted in split hand/split foot protein 1, deleted in split-hand/split-foot 1, DSS1Split hand/foot malformation type 1 protein, ECD, SEM1, Shfdg1, SHSF1SHFD1, Split hand/foot deleted protein 1, split hand/foot malformation (ectrodactyly) type 1
Host Species Rabbit
Immunogen Recombinant protein of human SHFM1.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7979
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.