missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Mouse anti-Broad Species Reactivity, Clone: K2, Novus Biologicals™

Product Code. 18371062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371062 5 mL 5mL
18317583 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18371062 Supplier Novus Biologicals Supplier No. NBP3113985ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen dsRNA
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone K2
Conjugate Unconjugated
Dilution ELISA, Immunofluorescence
Formulation Culture supernatant (RPMI, 5% fetal calf serum)
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Purification Method Unpurified
Quantity 5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.