missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Mouse anti-Broad Species Reactivity, Clone: J2, Novus Biologicals™

Product Code. 18031536 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 ug
Unit Size:
200µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031536 200 μg 200µg
18328811 500 ug 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18031536 Supplier Novus Biologicals Supplier No. NBP311395200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification
TRUSTED_SUSTAINABILITY

Specifications

Antigen dsRNA
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification
Classification Monoclonal
Clone J2
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Immunoaffinity Purification
Formulation Lyophilised from PBS
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution The lyophilised sample should be reconstituted with 200 ul (500 ul for 500 ug size) sterile distilled water. The mAb will then be in PBS without any stabilisers at a concentration of 1 mg/ml.
Target Species Multi-species
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.