missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ dsRNA Marker Easy

Product Code. 16052038
Change view
Click to view available options
Quantity:
125 μL
Unit Size:
125µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16052038 125 μL 125µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16052038 Supplier Abnova™ Supplier No. R0006.125uL
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

an ideal size marker for determinating sizes of double-stranded RNAs

  • The dsRNA Marker Easy is supplied in a ready-to-load mixture of loading dye (containing Tris-HCl buffer, glycerol, EDTA sodium salt, sodium chloride, bromphenol blue) and an ideal size marker for determinating sizes of double-stranded RNAs.
  • The dsRNA Marker Easy consists of ten double-stranded RNAs, 10, 20, 30, 50, 100, 200, 300, 400, 500 and 1,000 base pairs.
  • In 5 ul of the dsRNA Marker Easy, a 20 bp of dsRNA is approximately 50 ng.
  • The dsRNA Marker Easy can be visualized by UV light after ethidium bromide staining.

Specifications

For Use With (Application) Electrophoresis
Quantity 125 μL
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.