missing translation for 'onlineSavingsMsg'
Learn More

DSPG3/EPYC Antibody, Biotin, Novus Biologicals™

Product Code. 18386954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386954 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386954 Supplier Novus Biologicals Supplier No. NBP300204B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DSPG3/EPYC Polyclonal specifically detects DSPG3/EPYC in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSPG3/EPYC
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, Pg-Lb, PGLB, Proteoglycan-Lb, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant DSPG3/EPYC (AAH30958.1; Met1-Val322).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1833
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.