missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DSPE-PEG 2000 Maleimide

Product Code. 30000436
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000436 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000436 Supplier Tocris Bioscience™ Supplier No. 7947/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PEGylated DSPE with a maleimide reactive group

DSPE-PEG (2000) maleimide is a functionalized phospholipid-polymer conjugate. It comprises a 1,2-distearoyl-sn-glycero-PE (DSPE) covalently coupled to a linear polyethylene glycol (Mr 2000) with a maleimide-terminated end. It is used to synthesize antibody-coupled PEG (2000)-DSPE micelles that can be transferred into liposomes to generate immunoliposomes. DSPE-PEG (2000) maleimide can also be incorporated into the outer membrane of lipid nanoparticles (LNPs) for further bioconjugation. It is used in antibody-targeted LNP-mRNA systems to generate transient antifibrotic chimeric antigen receptor (CAR) T cells in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine-N-[maleimide(polyethylene glycol-2000)]
Quantity 10 mg
Target Lipid Nanoparticles
Molecular Formula (C2H4O)nC51H91N3O13P.NH4 or Na
Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.