missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DSPE-PEG 2000 Biotin
Description
DSPE-PEG 2000 Biotin is a biotinylated phospholipid conjugate designed for applications in lipid-based drug delivery, bioconjugation, and targeted molecular interactions. DSPE-PEG 2000 Biotin comprises 1,2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE), a polyethylene glycol (PEG 2000) spacer, and a biotin functional group enabling biotin-streptavidin binding.
Specifications
Specifications
| Chemical Name or Material | 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] |
| CAS | 474964-24-4 |
| Quantity | 10 mg |
| Target | Lipid Nanoparticles |
| Molecular Formula | (C2H4O)nC54H100N4O12PS.NH4 |
| Purity | 0.9 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?