missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DSPE-PEG 2000 Azide
Description
DSPE-PEG 2000 Azide is a functionalized phospholipid-polymer conjugate. It comprises a 1,2-distearoyl-sn-glycero-PE (DSPE) covalently coupled to a linear polyethylene glycol (Mr 2000) with an azide-terminated end for onward chemistry. It is commonly used to synthesize antibody-coupled liposomes and lipid nanoparticles. It has been used to make macrophage and EGFR antibody targeted LNPs using strain-promoted azide alkyne cycloaddition (SPAAC) click reaction.
Specifications
Specifications
| Chemical Name or Material | 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine-N-[carbonyl-azido(polyethyleneglycol)-2000] |
| CAS | 1938081-40-3 |
| Quantity | 10 mg |
| Target | Lipid Nanoparticles |
| Molecular Formula | (C2H4O)nC44H84N4O10P.NH4 |
| Purity | 0.9 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?