missing translation for 'onlineSavingsMsg'
Learn More

20(S)-Ginsenoside Rg3, MedChemExpress

Product Code. 16484857
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16484857

Brand: Medchem Express HYN0603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 20(S)-Ginsenoside Rg3 is the main component of Red ginseng. Ginsenoside Rg3 inhibits Na+ and hKv1.4 channel with IC50s of 32.2±4.5 and 32.6±2.2 μM, respectively. 20(S)-Ginsenoside Rg3 also inhibits Aβ levels, NF-κB activity, and COX-2 expression.

20(S)-Ginsenoside Rg3 is the main component of Red ginseng. Ginsenoside Rg3 inhibits Na+ and hKv1.4 channel with IC50s of 32.2±4.5 and 32.6±2.2 μM, respectively. 20(S)-Ginsenoside Rg3 also inhibits Aβ levels, NF-κB activity, and COX-2 expression.

Specifications

Chemical Name or Material 20(S)-Ginsenoside Rg3
CAS 14197-60-5
Color White
Purity Grade Notes Research
Molecular Formula C42H72O13
Quantity 50 mg
Synonym 20(S)-Propanaxadiol S-ginsenoside Rg3
Solubility Information DMSO : 100 mg/mL (127.39 mM; Need ultrasonic)
SMILES O[C@@H]([C@@H]([C@@H](CO)O1)O)[C@@H](O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)[C@@H]1O[C@H]3CC[C@]4(C)[C@@]5([H])C[C@@H](O)[C@]6([H])[C@@H](C(C)(O)CC/C=C(C)\C)CC[C@](C)6[C@@](C)5CCC4C3(C)C
Molecular Weight (g/mol) 785.01
Formula Weight 785.01
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.