missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSN1 Antibody (2A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079980-M01
This item is not returnable.
View return policy
Description
DSN1 Monoclonal antibody specifically detects DSN1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| DSN1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| C20orf172, chromosome 20 open reading frame 172, dJ469A13.2, DSN1, MIND kinetochore complex component, homolog (S. cerevisiae), FLJ13346, hKNL-3, kinetochore null 3 homolog, kinetochore-associated protein DSN1 homolog, KNL3, MIS13MGC32987 | |
| C20orf172 (NP_079194.2, 257 a.a. ∽ 355 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPAIHGSGSGSC | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 2A7 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_079194 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79980 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction