missing translation for 'onlineSavingsMsg'
Learn More

DSM265, MedChemExpress

Product Code. p-200098936
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489486

Brand: Medchem Express HY100184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DSM265 is a long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH) with an IC50 of 8.9 nM. DSM265 can also inhibit the growth of Pf3D7 parasites with an EC50 of 4.3 nM.

DSM265 is a long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH) with an IC50 of 8.9 nM. DSM265 can also inhibit the growth of Pf3D7 parasites with an EC50 of 4.3 nM.

Specifications

Chemical Name or Material DSM265
Physical Form Solid
CAS 1282041-94-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H12F7N5S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (120.39 mM; Need ultrasonic) ∣Ethanol : 11.11 mg/mL (26.75 mM; Need ultrasonic)
SMILES CC1=NC2=NC(C(C)(F)F)=NN2C(NC3=CC=C(S(F)(F)(F)(F)F)C=C3)=C1
Molecular Weight (g/mol) 415.33
Formula Weight 415.33
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.