missing translation for 'onlineSavingsMsg'
Learn More

DSM265, MedChemExpress

Product Code. 16489486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489486 50 mg 50mg
16462828 10 mg 10mg
16454246 100 mg 100mg
16412536 25 mg 25mg
16405577 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489486 Supplier Medchem Express Supplier No. HY100184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DSM265 is a long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH) with an IC50 of 8.9 nM. DSM265 can also inhibit the growth of Pf3D7 parasites with an EC50 of 4.3 nM.

DSM265 is a long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH) with an IC50 of 8.9 nM. DSM265 can also inhibit the growth of Pf3D7 parasites with an EC50 of 4.3 nM.

Specifications

Chemical Name or Material DSM265
Physical Form Solid
CAS 1282041-94-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H12F7N5S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (120.39 mM; Need ultrasonic) ∣Ethanol : 11.11 mg/mL (26.75 mM; Need ultrasonic)
SMILES CC1=NC2=NC(C(C)(F)F)=NN2C(NC3=CC=C(S(F)(F)(F)(F)F)C=C3)=C1
Molecular Weight (g/mol) 415.33
Formula Weight 415.33
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.