missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21880-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies. The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| DSG2 | |
| Polyclonal | |
| Unconjugated | |
| DSG2 | |
| ARVC10, ARVD10, Cadherin family member 5, CDHF5, desmoglein 2, DSG2, HDGC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13511, 1829, 307562 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O55111, Q14126 | |
| Dsg2 | |
| DSG2 Fusion Protein Ag16393 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction