missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10452-1-AP-20UL
This item is not returnable.
View return policy
Description
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.Specifications
| DSE | |
| Polyclonal | |
| Unconjugated | |
| DSE | |
| dermatan sulfate epimerase, DS epimerase, DSE, DSEPI, SART 2, SART2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29940 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UL01 | |
| DSE | |
| DSE Fusion Protein Ag0695 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction