missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsDNA Mouse anti-Human, Clone: rDSD/4565, Novus Biologicals™
Mouse Monoclonal Antibody
£219.00
Specifications
| Antigen | dsDNA |
|---|---|
| Clone | rDSD/4565 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
dsDNA Monoclonal specifically detects dsDNA in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| dsDNA | |
| 0.2 mg/ml | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG2a κ | |
| Bioreactor concentrate |
| rDSD/4565 | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Mouse | |
| Human | |
| Nuclei of Burkitt's cells | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title