missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsDNA Antibody (rDSD/8266) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24019JF585
This item is not returnable.
View return policy
Description
dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| dsDNA | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Double stranded DNA | |
| Nuclei of Burkitt's cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rDSD/8266 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction