missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DSCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564995
This item is not returnable.
View return policy
Description
Immunogen sequence: KLLNHVTQLVD SESWSFGKVP LNTCLQELGP LEPEEMIEHC LKCYGKKYVD EGEVYFELDA DKICRAAARM LLQNAVKFNL AEFQEVWQ Highest antigen sequence identity to the following orthologs - mouse 87%, rat 92%.
CHTF18 (MIM 613201), CHTF8 (MIM 613202), and DSCC1 arecomponents of an alternative replication factor C (RFC) (see MIM600404) complex that loads PCNA (MIM 176740) onto DNA during Sphase of the cell cycle.
Specifications
| DSCC1 | |
| Polyclonal | |
| Unconjugated | |
| DSCC1 | |
| 2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79075 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BVC3 | |
| DSCC1 | |
| Recombinant Human DSCC1. Recombinant protein control fragment (Product #RP-104575). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction