Learn More
Description
The DS-36 Matrix Standard (Dye Set J6) is used to perform the spectral calibration required to analyze 6-FAM, VIC, NED, SID, TAZ, and LIZ dye-labeled DNA fragments on the Applied Biosystems SeqStudio, 3130, 3500, and 3730 series systems. The data collection software for these instruments uses the multi-component matrix to automatically analyze the 6 differently colored fluorescent dye-labeled samples in a single capillary. The DS-36 Matrix Standard (Dye Set J6) contains 6 specific sizes of a unique fluorescent dye label.
Specifications
Specifications
| Software Type | Data Collection Software |
| Starting Material | DNA |
| Quantity | 8 Runs |
| For Use With (Equipment) | 3500xL Genetic Analyzer, 3130xl Genetic Analyzer, 3130 Genetic Analyzer, 3500 Genetic Analyzer |
| Label or Dye | 6-FAM, LIZ, NED, SID, TAZ, VIC |
| Product Type | DS-36 Matrix Standard |
| Shipping Condition | Wet Ice |
| Content And Storage | Store in refrigerator (2–8°C). |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.