missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-33 Matrix Standard Kit (Dye Set G5)

Product Code. 15949182 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
8 Runs
Unit Size:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15949182 8 Runs 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949182 Supplier Applied Biosystems™ Supplier No. 4345833

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains 5 dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution

Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples.

The Matrix Standard Kit is used to generate the multicomponent matrix required when analyzing 6-FAM, VIC, NED, PET, and LIZ-labeled DNA fragments on the Applied Biosystems SeqStudio, 3500, and 3730 series Systems. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples in a single capillary.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition Wet Ice
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer; SeqStudio
Quantity 8 Runs

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.