missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-33 GeneScan™ Installation Standards with GeneScan™ 600 LIZ™ Size Standard v2.0

Product Code. 15809826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809826 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809826 Supplier Applied Biosystems™ Supplier No. 4376911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Each kit contains four tubes of GeneScan™ Installation Standard DS-33 and one tube of GeneScan™ 600 LIZ™ size standard v2.0.

The DS-33 GeneScan™ Installation standard consists of pooled PCR products labeled with 6-FAM™, VIC™, NED™ and PET™ dyes.

The kit also contains a tube of GeneScan™ 600 LIZ™ Size Standard 2.0.

Order Info

Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each kit contains four tubes of GeneScan™ Installation Standard DS-33 and one tube of GeneScan™ 600 LIZ™ size standard v2.0.
Store the DS-33 tubes at -25° to -15°C
Store the GeneScan 600 LIZ Size Standard v2.0 opon thawing at 2@deg; to 8°C, protect from light.
Format Tube
For Use With (Equipment) 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer
Label or Dye FAM, LIZ, NED, PET, VIC
Product Line GeneScan, LIZ
Product Type Installation Standard
Quantity 1 kit
Shipping Condition Dry Ice
Test Time Standard

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.