missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-31 Matrix Standard Kit (Dye Set D)

Product Code. 15889796 Shop All Applied Biosystems Products
Click to view available options
Quantity:
8 runs
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15889796

Brand: Applied Biosystems™ 4345829

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Contains 4 oligonucleotides labeled respectively with 6FAM™,VIC™,NED™,and ROX™

Suitable for Dye Set D spectral calibration, on ABI PRISM™ Analyzers: 3100-Avant™, 3100, and 3730. Supports 4-Dye fragment analysis applications.

This kit contains 4 oligonucleotides respectively labeled with 6-FAM™, VIC™, NED™, and ROX™, pre-pooled in a single tube. Suitable for Dye Set D spectral calibration on Applied Biosystems Genetic Analyzers. Supports 4-Dye fragment analysis applications (Linkage Mapping Set v2.5, Mouse Mapping Primers).

The DS-31 Matrix Standard Set is used to generate the “multicomponent matrix” required when analyzing labeled DNA fragments on Applied Biosystems Genetic Analyzers. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition Wet Ice
For Use With (Equipment) 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer
Quantity 8 runs

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.