missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-30 Matrix Standard Kit (Dye Set D)

Product Code. 15899712 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
8 runs
Unit Size:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899712 8 runs 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15899712 Supplier Applied Biosystems™ Supplier No. 4345827
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Contains 4 oligonucleotides labeled respectively with 6FAM™,HEX™,NED™,and ROX™

This Matrix Standard Set is used to generate the 'multicomponent matrix' required when analyzing fluorescent dye-labeled DNA fragments.

The DS-30 Matrix Standard kit contains 4 oligonucleotides respectively labeled with 6-FAM™, HEX™, NED™, and ROX™, pre-pooled in a single tube. This Matrix Standard Set is used to generate the “multicomponent matrix” required when analyzing fluorescent dye-labeled DNA fragments on the Applied Biosystems™ Systems. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition Wet Ice
For Use With (Equipment) SeqStudio 3500 and 3730xl
Quantity 8 runs

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.