missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-02 Matrix Standard Kit, for 3500/3730/SeqStudio™/SeqStudio™ Flex

Product Code. 15859756 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
8 Runs
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859756 8 Runs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15859756 Supplier Applied Biosystems™ Supplier No. 4323014
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Contains dye-labeled oligonucleotides used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit

The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA™, dROX™, and LIZ™) are used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit.

The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA, dROX, and LIZ) are used to generate the 'multicomponent matrix' required for the SNaPshot Multiplex Kit. The Data Collection software utilizes the multicomponent matrix to automatically correct for the spectral overlap in samples labeled with DS-02 dyes. The kit consists of one tube of matrix standard which is sufficient for a minimum of eight array runs (on a 24 capillary) or two array runs (on a 96 capillary).

Matrix standards do not need to be run with every set of sample injections. The standard only needs to be run once in order to generate a matrix file which is then applied to samples run under similar conditions. For more information on the use of matrix standards, refer to the instrument User Manual or Getting Started Guide.

Used with SNaPshot Multiplex Kit on 3500/3500xl, 3730/3730xl, SeqStudio, and SeqStudio Flex series genetic analyzers.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains dye-labeled oligonucleotides used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit. Store at 2 - 8°C. Do not freeze.
Format Kit
For Use With (Equipment) SeqStudio™ Genetic Analyzer, 3730 Series Genetic Analyzers, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye LIZ, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA
Product Type DS-02 Matrix Standard Kit
Quantity 8 Runs
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.