missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Dry-Block Heater/Cooler
Click to view available options
Tube Size:
1.5 mL
2 mL
Unit Size:
Each
Description
The PCH cooler/heaters enable a stable temperature to be maintained during room temperature fluctuations.
- The ingenious block construction, combined with a powerful Peltier module, produces very rapid cooling and heating for swift change of application and increased flexibility. In addition, the Peltier effect produces efficient cooling and heating for small thermal loads without the use of a refrigeration system.
- The dry temperature control system maintains a clean working environment, reducing the chance of contamination or sample dilution. A Peltier system, without the vibration of refrigeration or the turbulence of a waterbath, is also ideal for all incubations where stable conditions are critical.
- The PCH is a very effective tool for DNA sample preparation for denaturing electrophoresis. The exceptionally rapid heat up enables the denaturation temperature of 95°C to be reached swiftly, the timer will signify denaturation time-up, and the samples can be rapidly cooled again ready to load the gels.
- Cooling/heating setting range from – 10ºC to 100°C, with very rapid cool down and heat-up times
- Stability ± 0.1°C, uniformity ±0.25°C at 37°C
- Choice of two models; capacity for up to 32 microtubes in a combination of two sizes: 12 x 1.5 mL plus 20 x 0.5 mL (PCH-1) or up to 20 microtubes of one size: 20 x 1.5 mL tubes (PCH-2), up to a total of 20 x 2 mL microtubes (PCH-3)
- Convenient integral reaction timer with audible alarm
- Dry temperature control system maintains a clean and vibration-free working environment
- A 2-line LCD display clearly indicates both set and actual values for temperature and time. Works in combination with simple push buttons for easy set-up.
- Flexible tube sizes and rapid heat-up/cool-down times enables swift change of application
Applications:
Life science – storing frozen restriction enzymes, nick translations, ligation reactions, restriction digests, protein solubilisation for PAGE, warm incubation of microcentrifuge tubes for hybridisation, enzyme reactions and deactivations.
Clinical – cooling blood samples prior to coagulation testing.
Specifications
Specifications
| Color | White |
| Description | Dry-block heater/cooler for 20 x 2.0 mL microtubes -10°C to +100°C (ambient -30°C minimum) |
| Holds | 20 Microtubes |
| Plug Type | US, EU, UK, AUS Plug |
| Temperature Range (Metric) | -10°C to +100°C (Cooling/Heating Setting Range) |
| Type | Dry-Block Heater/Cooler |
| Display | 2 line x 16 character LCD |
| Temperature Stability | Stability ±0.1°C, uniformity ±0.25°C at 37°C |
| Timer | 1 min. to 96 hr., Non-stop |
| Tube Size | 2 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction