missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Digital block heater for four aluminum blocks
- Included PT-1000 temperature sensor allows temperature control directly in the sample vessel
- Block heater is ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood-urea-nitrogen determinations and in situ hybridizations
- Stepless adjustable temperature up to 120°C
- Timer: Countdown, adjustable from 1 min. to 99h 59 min.
- Counter: Displays heating time
- Highly versatile with a large number of blocks
- Error code display
- Fixed safety circuit
- Acoustic signal at end of test
Specifications
Specifications
| Temperature Range (Metric) | Ambient to 120°C |
| Type | Dry Block Heater |
| No. of Blocks | 4 |
| Width (Metric) | 151 mm |
| Height (Metric) | 73 mm |
| Electrical Requirements | 220/240 V, 50/60 Hz |
| Voltage | 220/240 V |
| Frequency | 50/60 Hz |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
