Learn More
Thermo Scientific™ Dry-Bags™ Reusable Filter
Filter water coming into Thermo Scientific™ Dry-Bags™ with a reusable filter that can be used up to 10 times if autoclaved between each use.
Brand:  Thermo Scientific™ DB0200A
Additional Details : Weight : 0.01000kg
Product Code. 11489848
  • £78.55 / Each
In Stock
to see stock.
Add to basket
Description
Description
Remove the need to pre-sterilize water prior to filling Thermo Scientific™ Dry-Bags™ using a sterile, reusable, 0.2 micron Thermo Scientific Dry-Bags Reusable Filter.
Obtain up to 10 uses from the Dry-Bags Reusable Filter, if autoclaved at 121ºC for 15 minutes between each use.
Attatch the filter to the filling tube of any Dry-Bags product requiring a separate filter and secure it with tube ties provided in each Dry-Bags product package.
Dry-Bags Reusable Filters are able to withstand 2.5L/minute of deionized water. The number of fills and fill speed depends on water particulate levels.
Contact us to arrange a demonstration of the Peristaltic Pump and its accessories.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
Dry-Bags Reusable Filter
0.2 μm
Includes reusable filter only
0.050 kg
2.5 L/min.
Dry-Bags
Dry-Bags Reusable Filter