Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Product Code. 17378272 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
1.5/2.0 mL Tubes
10/15 mL and 5/7 mL Tubes
14 mm dia. Tubes
15 mL Tubes
17 mm dia. Tubes
18.5 mm dia. Tubes
26 mm dia. Tubes
30 mm dia. Tubes
50 mL Tubes
No. of Positions:
15
30
50
60
64
76
142
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. of Positions unitSize
17378272 1.5/2.0 mL Tubes 50 Each
17388272 15 mL Tubes 30 Each
17398272 50 mL Tubes 15 Each
17308282 10/15 mL and 5/7 mL Tubes 30 Each
17318282 14 mm dia. Tubes 64 Each
17328282 18.5 mm dia. Tubes 64 Each
17338282 17 mm dia. Tubes 142 Each
17348282 26 mm dia. Tubes 76 Each
17358282 30 mm dia. Tubes 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 17378272 Supplier Thermo Scientific™ Supplier No. 88882149
Only null left
Add to basket
Add to basket
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
TRUSTED_SUSTAINABILITY
Specifications
Holds 1.5/2.0 mL Tubes
Product Type Rotating Drum
Material Stainless Steel
Diameter (Metric) 290 mm
For Use With (Equipment) Accessory for Digital Cel-Gro Tissue Culture Rotator, 120V (88882015) and Digital Cel-Gro Tissue Culture Rotator, 230V (88882016)
No. of Positions 50