Learn More
Eppendorf™ Rotor T-60-11 with Lid
Product Code. 10104771 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
5810/5810R Centrifuge
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10104771 5810/5810R Centrifuge Each
1 options
Product Code. 10104771 Supplier Eppendorf™ Supplier No. 5804730003
Designed for use with Eppendorf Centrifuge 5810/5810 R
  • Rotor T-60-11
  • For Centrifuge 5810/5810 R
  • Includes rotor lid
  • For 1.5/2.0mL tubes
  • Without adapter
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor T-60-11 with Lid
For Use With (Equipment) 5810/5810R Centrifuge
Includes Lid