missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 (N-terminal) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26187-1-AP-150UL
This item is not returnable.
View return policy
Description
Dynamin-related protein 1 (DRP1) is a member of the dynamin superfamily of GTPases. Members of this family have diverse cellular functions including vesicle scission, organelle fission, viral resistance, and intracellular trafficking (reviewed in 1). DRP1 affects mitochondrial morphology and is important in mitochondrial and peroxisomal fission in mammalian cells.Specifications
| DRP1 (N-terminal) | |
| Polyclonal | |
| Unconjugated | |
| DNM1L | |
| DLP1, DNM1L, DRP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10059, 74006 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00429, Q8K1M6 | |
| Dnm1l | |
| DRP1 (N-terminal) Fusion Protein Ag24249 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction